missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G Sepharose™ 4 Fast Flow Media

Product Code. 11544935 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
Unit Size:
200mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544935 200 mL 200mL
11524935 5 mL 5mL
11534935 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11544935 Supplier Cytiva Supplier No. 17061805
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.

  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications

TRUSTED_SUSTAINABILITY

Specifications

Ligand Concentration 2mg Protein G/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 3 to 9
Quantity 200 mL
Content And Storage 2°C to 8°C
For Use With (Application) Used in a range of diagnostic and research applications
Ligand Recombinant Protein G lacking albumin binding region
compliance-icons
Product Identifier
  • Protein G Sepharose 4 Fast Flow, 200 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.