missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Disulfide Isomerase/P4HB Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £367.00
Specifications
| Antigen | Protein Disulfide Isomerase/P4HB |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312427
|
Novus Biologicals
NBP3-03678-20UL |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301016
|
Novus Biologicals
NBP3-03678-100UL |
100 μL |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Disulfide Isomerase/P4HB Polyclonal specifically detects Protein Disulfide Isomerase/P4HB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Protein Disulfide Isomerase/P4HB | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 329-508 of human Protein Disulfide Isomerase/P4HB (NP_000909.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia | |
| PBS with 50% glycerol, pH7.3. | |
| 5034 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title