missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Disulfide Isomerase/P4HB Rabbit anti-Human, Clone: 16, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89676-50UL
This item is not returnable.
View return policy
Description
Protein Disulfide Isomerase/P4HB Monoclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Protein Disulfide Isomerase/P4HB | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| 16 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 ul/mg of lysate | |
| Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55 | |
| Recombinant Human Protein Disulfide Isomerase/P4HB protein | |
| 50 μg | |
| Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia | |
| 5034 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction