missing translation for 'onlineSavingsMsg'
Learn More

Protein Disulfide Isomerase/P4HB Antibody (16), DyLight 405, Novus Biologicals™

Product Code. 18389730 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389730 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389730 Supplier Novus Biologicals Supplier No. NBP289676V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Protein Disulfide Isomerase/P4HB Monoclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Disulfide Isomerase/P4HB
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 16
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Protein Disulfide Isomerase/P4HB (NP_000909.2; Met 1-Lys 505).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 5034
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.