Learn More
Invitrogen™ Protein C Recombinant Mouse Monoclonal Antibody (HPC-4)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551846
Description
Specifically binds to a specific twelve peptide sequence (EDQVDPRLIDGK) in the activation region of the Protein C (spanning the thrombin cleavage site). Does not react with activated protein C. Has been shown to be cross-reactive with baboon Protein C but not with bovine or mouse Protein C.
Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
Specifications
| Protein C | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P04070 | |
| PROC | |
| This antibody was raised by immunising mice with human protein C. | |
| 200 μg | |
| Primary | |
| Human, Baboon | |
| Antibody | |
| IgG1 κ |
| Affinity Purification, ELISA, Inhibition Assays, Western Blot, Immunocytochemistry | |
| HPC-4 | |
| Unconjugated | |
| PROC | |
| Activation peptide; Anticoagulant protein C; APC; autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain | |
| Mouse | |
| Protein A | |
| RUO | |
| 5624 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.