Learn More
Protein C inhibitor Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10673-1-AP-150UL
 View more versions of this product
Product Code. 16867203
  • £349.00 / 150µL
Description
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
Protein C inhibitor
Polyclonal
Unconjugated
SERPINA5
PAI3, PCI, PLANH3, PROCI, Protein C inhibitor, SERPINA5
Rabbit
Antigen Affinity Chromatography
RUO
5104
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05154
SERPINA5
Protein C inhibitor Fusion Protein Ag1078
150 μL