missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Protein BlueRed Marker
Click to view available options
Quantity:
2 x 300 μL
Unit Size:
Pack of 2
Description
- The protein concentrations are optimized to give uniform band intensities.
- The maker is supplied in gel loading buffer for direct loading onto SDS-PAGE without heating or adding reducing agent.
Specifications
Specifications
| For Use With (Application) | WB,Electrophoresis |
| Quantity | 2 x 300 μL |
| Storage Requirements | Store at -20°C for 1 year Aliquot to avoid repeated freezing and thawing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction