missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A Sepharose™ CL-4B Medium

Product Code. 11564955 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1.5 g
25 mL
500 mL
Unit Size:
1500mg
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564955 25 mL 25mL
11554955 500 mL 500mL
11514945 1.5 g 1500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11564955 Supplier Cytiva Supplier No. 17096303

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For immunoglobin purification and fractionation. Cytiva Protein A Sepharose™ CL-4B Medium is also well suited for immunoprecipitation.

  • Strong binding to Fc region of IgG
  • Suitable for polyclonal antibody purification
  • High capacity—20mg human IgG/mL
  • pH stability Cleaning-in-Place (CIP): 2 to 10
  • Stable in commonly used aqueous buffers, 6M guanidine-HCl, 8M urea

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 20mg Human IgG/mL
Ligand Concentration 3mg Protein A/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 2 to 10
Quantity 25 mL
Content And Storage 2°C to 8°C
For Use With (Application) For immunoglobin purification and fractionation and is suitable for immunoprecipitation procedures
Ligand Native Protein A
compliance-icons
Product Identifier
  • Protein A Sepharose CL-4B, 25 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.