missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Binds Fc portion of IgG antibodies from a wide range of species
- Binds 5mg human IgG/mL gel
- Eliminates the need for a range of immunoadsorbent columns
- Column may be regenerated between separations by washing with low-pH buffer
Order Info
1mg Protein A/mL gel
Specifications
Specifications
| Binding Capacity | 5mg Human IgG/mL |
| Quantity | 10 mL |
| Content And Storage | 4°C to 30°C |
| For Use With (Application) | For separation of cells coated with antibodies |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?