missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A and Protein G Mag Sepharose™

Product Code. 10300695 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 μL
4 x 500 μL
Binding Capacity:
13 to 22mg Human IgG/mL
8 to 17mg Human IgG/mL
Ligand:
Native Protein A
Protein G
Bead Type:
Protein A Mag Sepharose
Protein G Mag Sepharose
Unit Size:
500µL
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Binding Capacity Ligand Bead Type unitSize
10300695 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose 500µL
10249574 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose 500µL
10488634 4 x 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose Pack of 4
10259574 4 x 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10300695 Supplier Cytiva Supplier No. 28944006
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Simplify enrichment of target proteins by immunoprecipitation or pull-down applications

Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

  • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: Native protein A; Native protein G
Binding capacity Protein A: 8-17mg human IgG/mL gel
Binding capacity Protein G: 13-22mg human IgG/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% ethanol
Storage temperature: 4°C to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Protein A Mag Sepharose
Binding Capacity 8 to 17mg Human IgG/mL
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Quantity 500 μL
Content And Storage 4°C to 8°C
For Use With (Application) Designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications, for biomarker research
Ligand Native Protein A
compliance-icons
Product Identifier
  • Protein A Mag Sepharose, 1 x 500 ul
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.