missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein A Polyclonal Antibody
Description
PA1-27415 detects protein A.
Protein A is an Orphan-U GPCR with an unknown ligand. This gene was identified from genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be expressed mainly in the brain. However, its function is not known. Three alternatively spliced variants have been described; the full length of the third variant is yet to be determined.
Specifications
Specifications
| Antigen | Protein A |
| Applications | ELISA, Lateral Flow, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Chicken |
| Immunogen | Purified recombinant protein A. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?