missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein A Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143390
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: 0.1 ng protein A.
Protein A is an Orphan-U GPCR with an unknown ligand. This gene was identified from genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be expressed mainly in the brain. However, its function is not known. Three alternatively spliced variants have been described; the full length of the third variant is yet to be determined.
Specifications
| Protein A | |
| Polyclonal | |
| Unconjugated | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgY |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Protein A. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction