missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A Mag Sepharose™ Xtra

Product Code. 11525664 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
Unit Size:
5mL
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11525664 2 x 1 mL Pack of 2
10565210 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11525664 Supplier Cytiva Supplier No. 28967056
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Magnetic beads are designed for rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants

Used as a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel and can be easily screen a larger number of samples in parallel with high throughput on a robotic device.

  • Visible and dense Sepharose-based magnetic beads with protein A ligand allow easy spotting and collection of bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Provides high purity and yield

∽350μg purified human IgG can be obtained from each purification run
Provides robust parallel screening of antibodies with high reproducibility
Matrix: paramagnetic, spherical, highly cross-linked agarose particles
Ligand: native protein A
Binding capacity: 27mg human IgG/mL gel
Operating temp.: room temperature
Stability: stable

  • Small-scale purification
  • Antibody screening
  • Antibody purification

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 27mg human IgG/mL gel
Quantity 2 x 1 mL
compliance-icons
Product Identifier
  • Protein A Mag Sepharose Xtra, 2 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.