missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Protective Jumpsuit with Hood, White
Description
- Protective and disposable
- Suitable for catering, food industry, industrial cleaning, laboratory, medical, and pharmaceutical applications
- Lightweight
- Breathable
- Opaque
- High-quality zipper
- Elasticized, ergonomic hood
- Individually packed
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Certifications/Compliance (Europe) | EC Category PPE: 0 |
| Weight (Metric) | 40 g/m2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?