missing translation for 'onlineSavingsMsg'
Learn More

EKASTU Safety Protective Hood 230

Product Code. 10310591
Change view
Click to view available options
Protective Hood 230
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10310591 Protective Hood 230
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10310591 Supplier EKASTU Safety Supplier No. 292082
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EKASTU Protective Hood 230 encloses the complete head, face and neck area.

  • Dust-proof and breathable fleece material
  • Large window shield made of triacetate
  • Limited use
  • 10 pieces per pack

Specifications

Color White
Product Type Respirator Hood
For Use With (Equipment) Particle Filter, Gas Filter, Multi-type Filter, Combined Filter, Multi-type Combined Filter
Dimensions (L x W x H) 21 x 26 x 3.5 cm
Weight (Metric) Approx. 295 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.