Learn More
Honeywell™ PROTECT (i)XTREM S3 Shoes
Product Code. 15816585
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15816585 37 1 pair
15826585 38 1 pair
15836585 39 1 pair
15846585 40 1 pair
15856585 41 1 pair
15866585 42 1 pair
15876585 43 1 pair
15886585 44 1 pair
15896585 45 1 pair
15806595 46 1 pair
15816595 47 1 pair
15826595 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15816585 Supplier Honeywell™ Supplier No. 624620437/7
Only null left
Add to basket
Add to basket
Honeywell™ PROTECT (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
  • Water resistant grain full leather
  • Ergonomic collar and padded tongue for an optimized comfort
  • Gusset to avoid water and dust penetration
  • Breathable 3D lining
  • Derby style for a better fitting in all sizes
  • Toe cap metal free
  • Metal free antiperforation insert
  • Injected PU / Nitrile sole
  • (i)NTELLIGENT wear control
  • EN ISO 20345:2011 S3 HRO HI CI SRC
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN ISO 20345:2011 S3 HRO HI CI SRC
Color Black
Material Leather
Size 37
Sole Material Polyurethane and nitrile
Toe Style Metal-free