missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 26S S5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99617-100ul
This item is not returnable.
View return policy
Description
Proteasome 26S S5 Polyclonal antibody specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot
Specifications
| Proteasome 26S S5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Proteasome 26S S5. | |
| 100 μg | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5711 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction