missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Proteasome 26S S5 Polyclonal antibody specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Proteasome 26S S5 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Proteasome 26S S5. |
| Quantity | 0.1 mL |
| Research Discipline | Neuroscience, Ubiquitin Proteasome Pathway |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?