missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 26S S5 Antibody (1H7), Novus Biologicals™

Product Code. 18386838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386838 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386838 Supplier Novus Biologicals Supplier No. H00005711M17

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteasome 26S S5 Monoclonal antibody specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 26S S5
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14478
Gene Alias KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic
Host Species Mouse
Immunogen PSMD5 (AAH14478.1, 1 a.a. ~ 504 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAQALALLREVARLEAPLEELRALHSVLQAVPLNELRQQAAELRLGPLFSLLNENHREKTTLCVSILERLLQAMEPVHVARNLRVDLQRGLIHPDDSVKILTLSQIGRIVENSDAVTEILNNAELLKQIVYCIGGENLSVAKAAIKSLSRISLTQAGLEALFESNLLDDLKSVMKTNDIVRYRVYELIIEISSVSPESLNYCTTSGLVTQLLRELTGEDVLVRATCIEMVTSLAYTHHGRQYLAQEGVIDQISNIIVGADSDPFSSFYLPGFVKFFGNLAVMDSPQQICERYPIFVEKVFEMIESQDPTMIGVAVDTVGILGSNVEGKQVLQKTGTRFERLLMRIGHQSKNAPVELKIRCLDAISSLLYLPPEQQTDDLLRMTESWFSSLSRDPLELFRGISSQPFPELHCAALKVFTAIANQPWAQKLMFNSPGFVEYVVDRSVEHDKASKDAKYELVKALANSKTIAEIFGNPNYLRLRTYLSEGPYYVKPVSTTAVEGAE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5711
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.