missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S beta2 Antibody (M1), Novus Biologicals™

Product Code. 18339508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339508 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339508 Supplier Novus Biologicals Supplier No. H00005690M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteasome 20S beta2 Monoclonal antibody specifically detects Proteasome 20S beta2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 20S beta2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone M1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. BC000268
Gene Alias EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2
Host Species Mouse
Immunogen PSMB2 (AAH00268, 1 a.a. ~ 201 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEYLIGIQGPDYVLVASDRVAASNIVQMKDDHDKMFKMSEKILLLCVGEAGDTVQFAEYIQKNVQLYKMRNGYELSPTAAANFTRRNLADCLRSRTPYHVNLLLAGYDEHEGPALYYMDYLAALAKAPFAAHGYGAFLTLSILDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLDNISFPKQGS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 5690
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.