missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S beta 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen Proteasome 20S beta 3
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229291
View Documents
Novus Biologicals
NBP3-38629-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30231660
View Documents
Novus Biologicals
NBP3-38629-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proteasome 20S beta 3 Polyclonal antibody specifically detects Proteasome 20S beta 3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

Proteasome 20S beta 3
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
EC 3.4.25.1, HC10-II, MGC4147, proteasome (prosome, macropain) subunit, beta type, 3, Proteasome chain 13, Proteasome component C10-II, proteasome subunit beta type-3, Proteasome theta chain
Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human Proteasome 20S beta 3 (NP_002786.2).,, Sequence:, MSIMSYNGGAVMAMKGKNCVAIAADRRFGIQAQMVTTDFQKIFPMGDRLYIGLAGLATDVQTVAQRLKFRLNLYELKEGRQIKPYTLMSMVANLLYEKRFGPYYTEPVIAGLDPKTFKPFICSLDLIGCPMVTDDFVVSGTCAEQMYGMCESLWEPNMDPDHLFETISQAMLNAVDRDAVSGMGVIVHIIEKDKITTRTLKARMD
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
5691
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.