missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S alpha 5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB120-11437-0.025mg

 View more versions of this product

Product Code. 18528950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Proteasome 20S alpha 5 Polyclonal specifically detects Proteasome 20S alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Proteasome 20S alpha 5
Polyclonal
Unconjugated
PBS containing 1mg/ml BSA with 0.05% Sodium Azide
PSMA5
Synthetic peptide corresponding to residues C A(120) L Q F G E E D A D P G A M(133) of human proteasome 20S subunit alpha 5.
0.025 mg
Core ESC Like Genes, Stem Cell Markers
5686
Human
IgG
Western Blot
1 mg/mL
Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500
EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA
Rabbit
Affinity Purified
RUO
Primary
This detects proteasome 20S subunit alpha 5 from human, rat and mouse cell lysates.
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only