missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S alpha 5 Antibody (AH1.1), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18429845 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429845 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429845 Supplier Novus Biologicals Supplier No. NBP276657MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteasome 20S alpha 5 Monoclonal antibody specifically detects Proteasome 20S alpha 5 in Human, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 20S alpha 5
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone AH1.1
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA
Host Species Mouse
Immunogen Sub-unit 5a
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5686
Target Species Human, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.