missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S alpha 5 Antibody (AH1.1), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18362610 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362610 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362610 Supplier Novus Biologicals Supplier No. NBP276657AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteasome 20S alpha 5 Monoclonal specifically detects Proteasome 20S alpha 5 in Human, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 20S alpha 5
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone AH1.1
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA
Gene Symbols PSMA5
Host Species Mouse
Immunogen Sub-unit 5a
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5686
Target Species Human, Xenopus species
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.