missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3322
This item is not returnable.
View return policy
Description
Proteasome 19S S7 Polyclonal specifically detects Proteasome 19S S7 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Proteasome 19S S7 | |
| Polyclonal | |
| Western Blot 1:1000, Immunoprecipitation 1:10 - 1:500 | |
| P35998 | |
| PSMC2 | |
| Human recombinant proteasome 19S subunit S7. | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Detects proteasome 19S subunit S7. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| 26S protease regulatory subunit 7, 26S proteasome AAA-ATPase subunit RPT1, MGC3004, MSS1ATPase, 2, proteasome (prosome, macropain) 26S subunit, ATPase, 2, Protein MSS1, putative protein product of Nbla10058 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5701 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction