missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S7 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97271AF405
This item is not returnable.
View return policy
Description
Proteasome 19S S7 Polyclonal specifically detects Proteasome 19S S7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Proteasome 19S S7 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| 26S protease regulatory subunit 7, 26S proteasome AAA-ATPase subunit RPT1, MGC3004, MSS1ATPase, 2, proteasome (prosome, macropain) 26S subunit, ATPase, 2, Protein MSS1, putative protein product of Nbla10058 | |
| Produced in rabbits immunized with E. coli-derived Human Proteasome 19S S7 fragment. | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5701 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction