missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Proteasome 19S 10B Recombinant Protein Antigen

Product Code. 18375856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375856 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375856 Supplier Novus Biologicals™ Supplier No. NBP317019PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 5708
Molecular Weight (g/mol) M.W. Theoretical: 28 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2
Gene Symbol PSMD2
Research Category Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Proteasome 19S 10B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.