missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 19S 10B Antibody (1B4), Novus Biologicals™

Product Code. 18355878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355878 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355878 Supplier Novus Biologicals Supplier No. H00005708M10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteasome 19S 10B Monoclonal antibody specifically detects Proteasome 19S 10B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 19S 10B
Applications ELISA
Classification Monoclonal
Clone 1B4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02368
Gene Alias 26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2
Host Species Mouse
Immunogen PSMD2 (AAH02368, 1 a.a. ~ 908 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEEGGRDKAPVQPQQSPAAAPGGTDEKPSGKERRDAGDKDKEQELSEEDKQLQDELEMLVERLGEKDTSLYRPALEELRRQIRSSTTSMTSVPKPLKFLRPHYGKLKEIYENMAPGENKRFAADIISVLAMTMSGERECLKYRLVGSQEELASWGHEYVRHLAGEVAKEWQELDDAEKVQREPLLTLVKEIVPYNMAHNAEHEACDLLMEIEQVDMLEKDIDENAYAKVCLYLTSCVNYVPEPENSALLRCALGVFRKFSRFPEALRLALMLNDMELVEDIFTSCKDVVVQKQMAFMLGRHGVFLELSEDVEEYEDLTEIMSNVQLNSNFLALARELDIMEPKVPDDIYKTHLENNRFGGSGSQVDSARMNLASSFVNGFVNAAFGQDKLLTDDGNKWLYKNKDHGMLSAAASLGMILLWDVDGGLTQIDKYLYSSEDYIKSGALLACGIVNSGVRNECDPALALLSDYVLHNSNTMRLGSIFGLGLAYAGSNREDVLTLLLPVMGDSKSSMEVAGVTALACGMIAVGSCNGDVTSTILQTIMEKSETELKDTYARWLPLGLGLNHLGKGEAIEAILAALEVVSEPFRSFANTLVDVCAYAGSGNVLKVQQLLHICSEHFDSKEKEEDKDKKEKKDKDKKEAPADMGAHQGVAVLGIALIAMGEEIGAEMALRTFGHLLRYGEPTLRRAVPLALALISVSNPRLNILDTLSKFSHDADPEVSYNSIFAMGMVGSGTNNARLAAMLRQLAQYHAKDPNNLFMVRLAQGLTHLGKGTLTLCPYHSDRQLMSQVAVAGLLTVLVSFLDVRNIILGKSHYVLYGLVAAMQPRMLVTFDEELRPLPVSVRVGQAVDVVGQAGKPKTITGFQTHTTPVLLAHGERAELATEEFLPVTPILEGFVILRKNPNYDL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5708
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.