Learn More
Thermo Scientific™ Protease Inhibitor Cocktail VII, for His-tag sequences, DMSO-free
Brand: Thermo Scientific™ J65553.LQ
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White | |
Air Sensitive | |
Lyophilized |
Protease Inhibitor Cocktail | |
-30°C to -10°C | |
1 Unit |