Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail VII
Product Code. 12801650
Change view
Click to view available options
:
Composition: AEBSF, Bestatin, E-64, Pepstatin A, Phosphoromidon
Product Code.
12801650 Composition: AEBSF, Bestatin, E-64, Pepstatin A, Phosphoromidon
1 options
Product Code. 12801650 Supplier Fisher BioReagents™ Supplier No. 12801650