Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail Vi For Plant Cell Extracts
Brand:  Fisher BioReagents™ 12891640
Code : 32
Additional Details : Weight : 0.10000kg
Product Code. 12891640
  • £70.23 / 1mL
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
Vial containing AEBSF HCl, Bestatin, E-64, Pepstatin A, Leupeptin, 1,10-Phenanthroline store in freezer at -20'C Fisher Bioreagents
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Protease Inhibitor Cocktail VI
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.