missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Protease Inhibitor Cocktail, mini-Tablet

Product Code. p-200104430
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Tablets
50 Tablets
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463826 25 Tablets Pack of 25
16414717 50 Tablets Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16463826 Supplier MedChemExpress Supplier No. HYK0011
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Protease inhibitor cocktail is used in cell lysates or tissue extracts to increase protein stability.

Specifications

Product Type Kits
Content And Storage Protease Inhibitor Cocktail (EDTA free, mini-Tablet) can be stored at 4°C for 2 years.
Shipping Condition Room temperature
Quantity 25 Tablets
Target VD/VDR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.