missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail I, Animal Free
Description
- Specificity: Serine Proteases, Esterases, Cysteine Proteases, Metalloproteases and Trypsin like Proteases
Specifications
Specifications
| Product Type | Protease Inhibitor Cocktail |
| Quantity | 1 Vial |
| Composition | AEBSF, Aprotinin (recombinant), E-64, EDTA, Leupeptin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?