missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protease-Activated Receptor-4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102297
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PAR4.
Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin.
Specifications
| Protease-Activated Receptor-4 | |
| Polyclonal | |
| Unconjugated | |
| F2RL3 | |
| coagulation factor II (thrombin) receptor-like 3; coagulation factor II receptor-like 3; F2R like thrombin/trypsin receptor 3; F2rl3; PAR4; PAR-4; protease-activated receptor 4; protease-activated receptor-4; proteinase-activated receptor 4; Thrombin receptor-like 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116498, 14065, 9002 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88634, Q920E0, Q96RI0 | |
| F2RL3 | |
| A synthesized peptide derived from human F2RL3(Accession Q96RI0), corresponding to amino acid residues G35-G85. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction