missing translation for 'onlineSavingsMsg'
Learn More

Protamine 1 Antibody (4F10), Novus Biologicals™

Product Code. 18320429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320429 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320429 Supplier Novus Biologicals Supplier No. H00005619M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protamine 1 Monoclonal antibody specifically detects Protamine 1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protamine 1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4F10
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH03673
Gene Alias cancer/testis antigen family 94, member 1, CT94.1, Cysteine-rich protamine, P1, protamine 1, sperm protamine P1, testis specific protamine 1
Host Species Mouse
Immunogen PRM1 (AAH03673.1, 1 a.a. ~ 51 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARYRCCRSQSRSRYYRQRQRSRRRRRRSCQTRRRAMRCCRPRYRPRCRRH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5619
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.