missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROTAC Sirt2 Degrader-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
PROTAC Sirt2 Degrader-1 is a SirReal-based PROTAC, acts as a Sirt2 degrader, composed of a highly potent and isotype-selective Sirt2 inhibitor, a linker, and a bona fide Cereblon ligand for E3 ubiquitin ligase. PROTAC Sirt2 Degrader-1 shows an IC50 of 0.25 μM for Sirt2, with no effect on Sirt1/Sirt3 (IC50s>100 μM).
Specifications
Specifications
| Chemical Name or Material | PROTAC Sirt2 Degrader-1 |
| CAS | 2098487-75-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C40H40N10O8S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (117.24 mM) |
| SMILES | O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(NCCCCN4C=C(COC5=CC(CC6=CN=C(NC(CSC7=NC(C)=CC(C)=N7)=O)S6)=CC=C5)N=N4)=O |
| Molecular Weight (g/mol) | 852.94 |
| Formula Weight | 852.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction