missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PROTAC™(H-PGDS)-8

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
PROTAC™(H-PGDS)-8 is the negative control for PROTAC™(H-PGDS)-7 (Cat. No. 8004).PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-[4-[4-[[4-[2,3-Dihydro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1,3-dioxo-1H-isoindol-4-yl]-1-piperazinyl]carbonyl]-1-piperidinyl]phenyl]-2-phenoxy-5-pyrimidinecarboxamide |
| CAS | 2761281-51-8 |
| Quantity | 2 mg |
| Target | Active Degrader Negative Controls |
| Molecular Formula | C41H40N8O7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction