missing translation for 'onlineSavingsMsg'
Learn More

PROTAC ERα Degrader-2, MedChemExpress

Product Code. 16474538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474538 10 mg 10mg
16427257 5 mg 5mg
16410847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474538 Supplier Medchem Express Supplier No. HY111846
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).

PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).

Specifications

Chemical Name or Material PROTAC ERα Degrader-2
CAS 1351169-29-3
Color White
Purity Grade Notes Research
Molecular Formula C42H61N5O8
Quantity 10 mg
Solubility Information DMSO : 200 mg/mL (261.79 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC/C2=N\OCC(NCCOCCOCCNC([C@H](CC(C)C)NC([C@@H](O)[C@H](N)CC3=CC=CC=C3)=O)=O)=O)([H])[C@@]4([H])[C@](C5=CC=C(O)C=C5CC4)([H])CC1
Molecular Weight (g/mol) 763.96
Formula Weight 763.96
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.