missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BET degrader-3, MedChemExpress

Product Code. 16493246
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493246 25 mg 25mg
16484798 10 mg 10mg
16447547 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493246 Supplier Medchem Express Supplier No. HY114229
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PROTAC BET Degrader-3 is a PROTAC connected by ligands for von Hippel-Lindau and BET.

PROTAC BET Degrader-3 is a PROTAC connected by ligands for von Hippel-Lindau and BET.

Specifications

Chemical Name or Material PROTAC BET degrader-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C53H64N12O9S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (95.67 mM; Need ultrasonic)
SMILES CCN1N=C(C2CC2)C=C1NC3=NC(C(NCCOCCOCC(N[C@@H](C(C)(C)C)C(N4[C@H](C(NCC5=CC=C(C6=C(C)N=CS6)C=C5)=O)C[C@@H](O)C4)=O)=O)=O)=NC7=C3C8=CC(OC)=C(C(C(C)=NO9)=C9C)C=C8N7
Molecular Weight (g/mol) 1045.22
Formula Weight 1045.22
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.