missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BET Degrader-1, MedChemExpress

Product Code. 16496597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496597 5 mg 5mg
16443878 10 mg 10mg
16442886 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496597 Supplier Medchem Express Supplier No. HY103633
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PROTAC BET Degrader-1 is a PROTAC connected by ligands for Cereblon and BET, decreasing BRD2, BRD3, and BRD4 protein levels at low concentration.

PROTAC BET Degrader-1 is a PROTAC connected by ligands for Cereblon and BET, decreasing BRD2, BRD3, and BRD4 protein levels at low concentration.

Specifications

Chemical Name or Material PROTAC BET Degrader-1
CAS 2093386-22-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C44H45N11O9
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (57.35 mM)
SMILES CCN1C(NC2=C3C4=CC(OC)=C(C(C(C)=NO5)=C5C)C=C4NC3=NC(C(NCCCCNC(COC6=C(C(N7C(CCC8=O)C(N8)=O)=O)C(C7=O)=CC=C6)=O)=O)=N2)=CC(C9CC9)=N1
Molecular Weight (g/mol) 871.9
Formula Weight 871.9
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.