missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BET-binding moiety 2, MedChemExpress

Product Code. 16480518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480518 5 mg 5mg
16428058 10 mg 10mg
16403977 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480518 Supplier Medchem Express Supplier No. HY43723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PROTAC BET-binding moiety 2 is an inhibitor of BET bromodomain.

PROTAC BET-binding moiety 2 is an inhibitor of BET bromodomain.

Specifications

Chemical Name or Material PROTAC BET-binding moiety 2
CAS 916493-82-8
Color White
Purity Grade Notes Research
Molecular Formula C20H17ClN4O4S
Quantity 5 mg
Solubility Information DMSO : 42.86 mg/mL (96.34 mM; Need ultrasonic)
SMILES CC1=C(C(O)=O)SC2=C1C(C3=CC=C(Cl)C=C3)=N[C@@H](CC(OC)=O)C4=NN=C(C)N24
Molecular Weight (g/mol) 444.89
Formula Weight 444.89
Percent Purity 98.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.