missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BET-binding moiety 1, MedChemExpress

Product Code. 16460487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460487 50 mg 50mg
16436837 5 mg 5mg
16424138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460487 Supplier Medchem Express Supplier No. HY107451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PROTAC BET-binding moiety 1 is a key intermediate for the synthesis of high-affinity BET inhibitors.

PROTAC BET-binding moiety 1 is a key intermediate for the synthesis of high-affinity BET inhibitors.

Specifications

Chemical Name or Material PROTAC BET-binding moiety 1
CAS 2093387-77-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H25N7O4
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (102.56 mM; Need ultrasonic)
SMILES O=C(C1=NC(NC2=CC(C3CC3)=NN2CC)=C4C(NC5=C4C=C(OC)C(C6=C(C)ON=C6C)=C5)=N1)O
Molecular Weight (g/mol) 487.51
Formula Weight 487.51
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.