missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostatic Acid Phosphatase/ACPP Antibody (SPM312), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Prostatic Acid Phosphatase/ACPP Monoclonal antibody specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Prostatic Acid Phosphatase/ACPP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SPM312 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase |
| Host Species | Mouse |
| Immunogen | Prostatic Acid Phosphatase/ACPP Purified from human seminal plasma (Uniprot: P15309) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?