missing translation for 'onlineSavingsMsg'
Learn More

Prostatic Acid Phosphatase/ACPP Antibody (ACPP/1339), DyLight 350, Novus Biologicals™

Product Code. 18410053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410053 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410053 Supplier Novus Biologicals Supplier No. NBP254417UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prostatic Acid Phosphatase/ACPP Monoclonal antibody specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for ELISA, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostatic Acid Phosphatase/ACPP
Applications ELISA, CyTOF
Classification Monoclonal
Clone ACPP/1339
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
Host Species Mouse
Immunogen Recombinant full-length human Prostatic Acid Phosphatase/ACPP protein (Uniprot: P15309)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.