missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostate Specific Acid Phosphatase (PSAP) Mouse, Clone: rACPP/1338, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | Prostate Specific Acid Phosphatase (PSAP) |
|---|---|
| Clone | rACPP/1338 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| Prostate Specific Acid Phosphatase (PSAP) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human ACPP protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 52kDa |
| rACPP/1338 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 55 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cytoplasmic | |
| PC12 cells or normal prostate or prostate carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title