missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Epredia™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-321-P0

 View more versions of this product

Product Code. 12640987

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Prostatic acid phosphatase (PSAP) is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase.

Host Species: Mouse

Clone: PASE/4LJ

Isotype: IgG1

Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.

Epitope: Not determined

Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma

Molecular Weight: 52kDa

Positive Control: Normal prostate or prostate CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prostate Specific Acid Phosphatase (PSAP) Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin)
PASE/4LJ
Unconjugated
Purified prostatic acid phosphatase (PSAP) from human seminal plasma
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.