missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostate Specific Acid Phosphatase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142740
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001127666.1; NP_001090.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.Specifications
Prostate Specific Acid Phosphatase | |
Polyclonal | |
Unconjugated | |
ACP3 | |
5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
55 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P15309 | |
ACP3 | |
Peptide with sequence C-NHMKRATQIPSYKK. | |
100 μg | |
Primary | |
Human | |
Liquid |