missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostate Specific Acid Phosphatase Monoclonal Antibody (PASE/4LJ), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513078
This item is not returnable.
View return policy
Description
MA5-13078 targets Prostate Specific Acid Phosphatase in IF and IHC (P) applications and shows reactivity with Human samples. The MA5-13078 immunogen is purified prostatic acid phosphatase (PSAP) from human seminal plasma.
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
Specifications
| Prostate Specific Acid Phosphatase | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P15309 | |
| ACP3 | |
| Purified prostatic acid phosphatase (PSAP) from human seminal plasma | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PASE/4LJ | |
| Unconjugated | |
| ACP3 | |
| 5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase | |
| Mouse | |
| Protein G | |
| RUO | |
| 55 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction