Learn More
Invitrogen™ Prostate Specific Acid Phosphatase Monoclonal Antibody (P1D8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514721
Description
MA5-14721 targets Prostatic Acid Phosphatase in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14721 immunogen is human Seminal Fluid PAP. MA5-14721 detects Prostatic Acid Phosphatase which has a predicted molecular weight of approximately 42 kDa. Product MA514721 is a smaller package size of MIP0101 (formerly sold as a Seradyn product).
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
Specifications
| Prostate Specific Acid Phosphatase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| P15309 | |
| ACP3 | |
| Human Seminal Fluid PAP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| P1D8 | |
| Unconjugated | |
| ACP3 | |
| 5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase | |
| Mouse | |
| Protein A | |
| RUO | |
| 55 | |
| Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.