missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostasin/Prss8 Rabbit anti-Human, Clone: 107, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89382-100UL
This item is not returnable.
View return policy
Description
Prostasin/Prss8 Monoclonal specifically detects Prostasin/Prss8 in Human samples. It is validated for ELISA.
Specifications
| Prostasin/Prss8 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5652 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 107 | |
| ELISA 1:1000-1:2000 | |
| CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, PROSTASIN, protease, serine, 8, Serine protease 8 | |
| Recombinant Human Prostasin/Prss8 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction